Human Elastase
Lieferant: Merck Millipore Calbiochem
324681-100
324681-50
324681-100EA
618
EUR
324681-100
324681-50
Human Elastase
Enzyme
Elastase, human neutrophil is a serine protease that degrades elastin, collagen, and proteoglycans. Its overexpression has been implicated in the development of pulmonary emphysema, rheumatoid arthritis, and cystic fibrosis.
- Source: prepared from blood that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV
- Specific activity: ≥20 units/mg protein
- Storage: –20 °C
Verpackung: Packed in glass bottle (100 μg) and plastic ampoule (50 μg).
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...