1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidin ≥95%, kristalliner Feststoff
Lieferant: Cayman Chemical
Gefahr
Synonyme: 1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine, Hhpp, 2,3,4,6,7,8-Hexahydro-1H-pyrimido[1,2-a]pyrimidine, TBD
20975-1 20975-10 20975-5
CAYM20975-1EA 43 EUR
CAYM20975-1 CAYM20975-10 CAYM20975-5
1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidin ≥95%, kristalliner Feststoff
1,5,7-Triazabicyclo[4.4.0]dec-5-ene
1,5,7-Triazabicyclo[4.4.0]dec-5-ene is a bicyclic guanidine base that can be used as a catalyst for a variety of base-mediated organic transformations, including Michael additions, Wittig reactions, Henry reactions, and transesterification reactions.
Formel: C₇H₁₃N₃
Molecular Weight: 139,20 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00043003
CAS-Nummer: 5807-14-7
UN: 1759
ADR: 8,II
Bestellen
Spezifikation Testergebnisse
Purity ≥95%