Total organic carbon standards, according to ISO/CEN EN1484, Supelco®
Lieferant: Merck
76067-250ML-F 76067-500ML-F
76067-250ML-FEA 114 EUR
76067-250ML-F 76067-500ML-F
Total organic carbon standards, according to ISO/CEN EN1484, Supelco®
Standards Umweltanalyse-Standards
  • Stabilized
  • Limited shelf life, expiry date on the label
Bitte beachten Storage temperature: 2…8 °C