(-)-Perillyl alcohol ≥96%, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-4-Isopropenyl-1-cyclohexenylmethanol, (S)-p-Mentha-1,8-dien-7-ol, (S)-(4-(Prop-1-en-2-yl)cyclohex-1-en-1-yl)methanol, (S)-(−)-Perillyl alcohol

M-Clarity™ Qualitätsstufe = MQ100

218391-10G 218391-50G
SIAL218391-10GEA 54.7 EUR
SIAL218391-10G SIAL218391-50G
(-)-Perillyl alcohol ≥96%, Sigma-Aldrich®
(-)-Perillyl alcohol
Formel: C₁₀H₁₆O
Molecular Weight: 152,24 g/mol
MDL: MFCD00062995
CAS-Nummer: 18457-55-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR