(2S)-Acetoxy(phenyl)essigsäure 99%

Lieferant: Thermo Scientific

Synonyme: (S)-2-Acetoxy-2-phenylacetic acid, (S)-(+)-α-Acetoxyphenylacetic Acid, (+)-O-Acetyl-L-mandelic Acid

H56567.03 H56567.06
H56567.03EA 36.8 EUR
H56567.03 H56567.06
(2S)-Acetoxy(phenyl)essigsäure 99%
(S)-(+)-O-Acetylmandelic acid
Formel: C₁₀H₁₀O₄
Molecular Weight: 194,19 g/mol
Schmelzpunkt: 97…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064215
CAS-Nummer: 7322-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR