(±)-2-Butanol ≥99,5% (durch GC), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
Lieferant: Honeywell Chemicals
|
Achtung
|
Synonyme:
(±)-Ethyl-methylcarbinol, (±)-Methyl-ethylcarbinol, (±)-sec-Butylalkohol, (±)-2-Hydroxybutan, (±)-sec-Butanol
19440-2.5L
19440-1L
19440-2.5LEA
175.12
EUR
19440-2.5L
19440-1L
(±)-2-Butanol ≥99,5% (durch GC), Puriss. p.a. Reag. Ph. Eur., Riedel-de Haën™
(±)-2-Butanol
Formel:
H₃CCH₂CH(OH)CH₃ Molecular Weight: 74,12 g/mol Siedepunkt: 99 °C Schmelzpunkt: –89 °C Dichte: 0,81 g/cm³ (20 °C) Flammpunkt: 24 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004569 CAS-Nummer: 78-92-2 EINECS: 201-158-5 UN: 1120 ADR: 3,III |
Spezifikation Testergebnisse
Refractive Index | n20/D 1.397 (lit.) |
Identity (NMR) | complying |
Assay (GC) | Min. 99.5 % |
Refractive index (n 20/D) | 1.396 - 1.398 |
Non-volatile matter | Max. 0.001 % |
Water (Karl Fischer) | Max. 0.1 % |
Aluminium (Al) | Max. 0.5 ppm |
Barium (Ba) | Max. 0.1 ppm |
Bismuth (Bi) | Max. 0.1 ppm |
Calcium (Ca) | Max. 0.5 ppm |
Cadmium (Cd) | Max. 0.05 ppm |
Cobalt (Co) | Max. 0.02 ppm |
Chromium (Cr) | Max. 0.02 ppm |
Copper (Cu) | Max. 0.02 ppm |
Iron (Fe) | Max. 0.1 ppm |
Potassium (K) | Max. 0.5 ppm |
Lithium (Li) | Max. 0.1 ppm |
Magnesium (Mg) | Max. 0.1 ppm |
Manganese (Mn) | Max. 0.02 ppm |
Molybdenum (Mo) | Max. 0.1 ppm |
Sodium (Na) | Max. 0.5 ppm |
Nickel (Ni) | Max. 0.02 ppm |
Lead (Pb) | Max. 0.1 ppm |
Strontium (Sr) | Max. 0.1 ppm |
Zinc (Zn) | Max. 0.1 ppm |
Aromatic substances (as C₆H₆) | Max. 0.01 % |
tert-Butanol | Max. 0.1 % |
Aldehydes (as HCHO) | Max. 100 ppm |
2-propanol (GC) | Max. 0.1 % |
methyl ethyl ketone (GC) | Max. 1000 ppm |
Ketones (as CH₃COCH₃) | Max. 100 ppm |
Vapor Density | 2.6 (vs air) |
Explosion Limit | 9.8 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...