3-Cyan-7-ethoxycumarin ≥97% (durch HPLC), Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ200
UC455-10MG
UC455-5MG
SIALUC455-10MGEA
567
EUR
SIALUC455-10MG
SIALUC455-5MG
3-Cyan-7-ethoxycumarin ≥97% (durch HPLC), Sigma-Aldrich®
3-Cyan-7-ethoxycumarin
Formel:
C₁₂H₉NO₃ Molecular Weight: 215,21 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00467359 CAS-Nummer: 117620-77-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...