Ammoniumnitrat ≥98%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™

Lieferant: Honeywell Chemicals
Achtung

Synonyme: Ammonsalpeter

31114.9050 31114-1KG 31114-6X1KG
HONE31114.9050EA 0 EUR
HONE31114.9050 31114-1KG 31114-6X1KG
Ammoniumnitrat ≥98%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Ammoniumnitrat
Formel: NH₄NO₃
Molecular Weight: 80,04 g/mol
Schmelzpunkt: 169 °C,(1.013 hPa).170 °C
Dichte: 1,72 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011425
CAS-Nummer: 6484-52-2
EINECS: 229-347-8
UN: 1942
ADR: 5.1,III
Merck Index: 13,00538

Bestellen

Spezifikation Testergebnisse

Assay Min. 98.0 %
Water insoluble matter Max. 0.005 %
Residue on ignition (SO₄) Max. 0.01 %
pH (5 %, 25 °C) 4.5 - 6.0
Calcium (Ca) Max. 0.002 %
Cadmium (Cd) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 1 ppm
Potassium (K) Max. 0.001 %
Magnesium (Mg) Max. 0.001 %
Sodium (Na) Max. 0.001 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 3 ppm
Nitrite (NO₂) Max. 0.0005 %
Phosphate (PO₄) Max. 5 ppm
Sulfate (SO₄) Max. 0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR