Eisen(II)chlorid Tetrahydrat ≥99,0% (RT), Puriss. p.a., Fluka™
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Eisenchlorür, Eisendichlorid Tetrahydrat
44939-250G
44939-1KG
44939-250GEA
67.43
EUR
44939-250G
44939-1KG
Eisen(II)chlorid Tetrahydrat ≥99,0% (RT), Puriss. p.a., Fluka™
Eisen(II)chlorid Tetrahydrat
Formel:
FeCl₂·4H₂O Molecular Weight: 198,81 g/mol Schmelzpunkt: 105…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149709 CAS-Nummer: 13478-10-9 UN: 3260 ADR: 8,III |
Spezifikation Testergebnisse
Assay | 99.0 - 101.0 % |
Arsenic (As) | Max. 5 ppm |
Calcium (Ca) | Max. 10 ppm |
Cadmium (Cd) | Max. 5 ppm |
Cobalt (Co) | Max. 50 ppm |
Chromium (Cr) | Max. 10 ppm |
Copper (Cu) | Max. 5 ppm |
Iron(III) | Max. 2000 ppm |
Potassium (K) | Max. 50 ppm |
Magnesium (Mg) | Max. 5 ppm |
Manganese (Mn) | Max. 50 ppm |
Sodium (Na) | Max. 50 ppm |
Nickel (Ni) | Max. 50 ppm |
Lead (Pb) | Max. 10 ppm |
Zinc (Zn) | Max. 20 ppm |
Sulfate (SO₄) | Max. 100 ppm |
Total nitrogen (as N) | Max. 10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...