Ammoniumfluorid ≥98%, Puriss. p.a. ACS, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Fluoroammonium

30101-50KG 30101.9050 30101-100G 30101-1KG 30101-6X1KG
30101-50KGEA 3147.03 EUR
30101-50KG HONE30101.9050 30101-100G 30101-1KG 30101-6X1KG
Ammoniumfluorid ≥98%, Puriss. p.a. ACS, Fluka™
Ammonium fluoride
Formel: NH₄F
Molecular Weight: 37,04 g/mol
Schmelzpunkt: 100 °C
Dichte: 1,01 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011423
CAS-Nummer: 12125-01-8
EINECS: 235-185-9
UN: 2505
ADR: 6.1,III
Merck Index: 13,00524

Bestellen

Spezifikation Testergebnisse

Assay Min. 98 %
Ammonium hydrogen difluoride (NH₄HF₂) Max. 1 %
Water insoluble matter Max. 0.005 %
Residue on ignition (SO₄) Max. 0.005 %
Cadmium (Cd) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0005 %
Potassium (K) Max. 0.002 %
Sodium (Na) Max. 0.002 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Pb) Max. 0.0005 %
Chloride (Cl) Max. 0.0005 %
Hexafluorosilicicate (as SiF₆) Max. 0.1 %
Sulfate (SO₄) Max. 0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR