Natriumhydroxid 98,0-100,5% (durch Acidimetrie) USP, NF, BP, Ph. Eur., pharmazeutischer Qualität

Lieferant: PanReac AppliChem

APPC141929.1214EA 110 EUR
APPC141929.1214 APPC141929.0416
Natriumhydroxid 98,0-100,5% (durch Acidimetrie) USP, NF, BP, Ph. Eur., pharmazeutischer Qualität
Sodium hydroxide
Formel: NaOH
Molecular Weight: 40 g/mol
Siedepunkt: 1,390 °C
Schmelzpunkt: 318 °C
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Assay (Acidim.) 98.0 - 100.5%
Assay (as Na) 54.0 - 59.8%
Identity according to Pharmacopoeias Passes Test
Insoluble matter in H₂O 0.025%
Insoluble Substances and Organic Matter Passes Test
Chloride (Cl) 0.0200%
Nitrogen Compounds (as N) 0.002%
Phosphate (PO₄) 0.002%
Sulfate (SO₄) 0.0200%
Carbonate (as Na₂CO₃) 1.0%
Residual solvents (Ph. Eur./USP) Passes Test
Fe 0.001%
K 0.5%
Elemental impurities (ICH Q3D), Class 1, Cd 0.5 ppm
Elemental impurities (ICH Q3D), Class 1, Pb 5 ppm
Elemental impurities (ICH Q3D), Class 1, As 1.5 ppm
Elemental impurities (ICH Q3D), Class 1, Hg 1.5 ppm
Elemental impurities (ICH Q3D), Class 2A, Co 5 ppm
Elemental impurities (ICH Q3D), Class 2A, V 10 ppm
Elemental impurities (ICH Q3D), Class 2A, Ni 20 ppm
Elemental impurities (ICH Q3D), Class 2B, Tl 5 ppm
Elemental impurities (ICH Q3D), Class 2B, Au 10 ppm
Elemental impurities (ICH Q3D), Class 2B, Pd 10 ppm
Elemental impurities (ICH Q3D), Class 2B, Ir 10 ppm
Elemental impurities (ICH Q3D), Class 2B, Os 10 ppm
Elemental impurities (ICH Q3D), Class 2B, Rh 10 ppm
Elemental impurities (ICH Q3D), Class 2B, Ru 10 ppm
Elemental impurities (ICH Q3D), Class 2B, Se 15 ppm
Elemental impurities (ICH Q3D), Class 2B, Ag 15 ppm
Elemental impurities (ICH Q3D), Class 2B, Pt 10 ppm
Elemental impurities (ICH Q3D), Class 3, Li 55 ppm
Elemental impurities (ICH Q3D), Class 3, Sb 120 ppm
Elemental impurities (ICH Q3D), Class 3, Ba 140 ppm
Elemental impurities (ICH Q3D), Class 3, Mo 25 ppm
Elemental impurities (ICH Q3D), Class 3, Cu 250 ppm
Elemental impurities (ICH Q3D), Class 3, Sn 600 ppm
Elemental impurities (ICH Q3D), Class 3, Cr 25 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR