AC1NUZLU (synthetisch)
Lieferant: BACHEM BIOCHEMICA
Synonyme:
Z-Val-Ala-DL-Asp-fluoromethylketone
4026865.0005
4026865.0025
BACE4026865.0005EA
0
EUR
BACE4026865.0005
BACE4026865.0025
AC1NUZLU (synthetisch)
AC1NUZLU
zVAD-FMK inhibited apoptosis induced by diverse stimuli. Preincubation of cells with this caspase-1 (ICE) inhibitor (50 µM) almost completely inhibited the formation of large kilobase-pair fragments of DNA, a characteristic biochemical change associated with apoptosis.
Formel:
C₂₁H₂₈FN₃O₇ Molecular Weight: 453,467 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD06795862 CAS-Nummer: 220644-02-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...