IGEPAL® CO-520, M.W. ~ 441, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Poly (oxyethylene)nonylphenyl ether), α-(Nonylphenyl)-ω-hydroxy-poly(oxy-1,2-ethanediyl) Branched Polymer

M-Clarity™ Qualitätsstufe = MQ100

238643-100G 238643-500G
SIAL238643-100GEA 96.5 EUR
SIAL238643-100G SIAL238643-500G
IGEPAL® CO-520, M.W. ~ 441, Sigma-Aldrich®
IGEPAL® CO-520

IGEPAL®CO-520 or poly (oxyethylene)nonylphenyl ether) is a non-ionic surfactant.


Application: Igepal aggregates have been used in fluorescence and light-scattering studies. Used in the preparation of monodispersed colloidal silica cadmium suphide nanocomposites, alumina nanoparticles, ultra thin silica coated magnetic nanoparticles.


IGEPAL is a registered trademark of Rhodia Operations.

Formel: (C₂H₄O)n·C₁₅H₂₄O
Dichte: 0,997 g/mL (25 °C) (Lit.)
MDL: MFCD00132851
CAS-Nummer: 68412-54-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR