Meclofenoxathydrochlorid 98%, Pulver

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(Dimethylamino)ethyl 2-(4-Chlorophenoxy)acetate Hydrochloride, 2-(4-Chlorophenoxy)acetic Acid 2-(Dimethylamino)ethyl Ester Hydrochloride, Centrophenoxine hydrochloride

J64196.06 J64196.14 J64196.22
J64196.06EA 38.3 EUR
J64196.06 J64196.14 J64196.22
Meclofenoxathydrochlorid 98%, Pulver
Meclofenoxathydrochlorid

Ester of dimethylethanolamine and 4-chlorophenoxyacetic acid.


Applications: Activates acetylcholinesterase activity in hippocampus of aged rats

Formel: C₁₂H₁₆ClNO₃
Molecular Weight: 294,17 g/mol
Schmelzpunkt: 135,..145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012533
CAS-Nummer: 3685-84-5
EINECS: 222-975-3
Merck Index: 14,05780

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR