D-Fructose-6-phosphate disodium salt, Hyrdrat ≥98%, amorphes Pulver

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ300

F3627-500MG F3627-100MG F3627-10MG F3627-1G F3627-5G
SIALF3627-500MGEA 122 EUR
SIALF3627-500MG SIALF3627-100MG SIALF3627-10MG SIALF3627-1G SIALF3627-5G
D-Fructose-6-phosphate disodium salt, Hyrdrat ≥98%, amorphes Pulver
D-Fructose-6-phosphate disodium salt

Fructose-6-phosphate is a glycolytic intermediate produced by the isomerization of glucose-6-phosphate by phosphoglucoisomerase.


D-Fructose 6-phosphate (F6P) is a sugar intermediate of the glycolytic pathway that may be used to help identify, differentiate and characterize enzymes such as phosphofructokinase(s), pyrophosphate-dependent fructose-6-phosphate 1-phosphotransferase(s), D-fructose-6-phosphate aldolase(s), glutamine:fructose-6-phosphate amidotransferase(s) and glucosamine-6P synthase(s).

Formel: C₆H₁₁Na₂O₉P
Lagertemperatur: Gefrierschrank
MDL: MFCD00135619
CAS-Nummer: 26177-86-6

Bestellen

Spezifikation Testergebnisse

Appearance (Color) White to Off-White
Appearance (Form) Powder
Solubility (Color) Colorless to Faint Yellow
Solubility (Turbidity) Clear to Slightly Hazy
Water ≤12%
Solvent, Alcohols ≤5%
Glucose-6-Phosphate ≤1.5 mole%
Fructose 1,6- diphosphate ≤0.05 mole%
% Phosphorus (Dry Basis) 9.9 - 10.8%
Sodium (Dry basis) 14.6 - 15.6%
% Purity (Dry basis) ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR