Glycine-N,N-bis(methylenephosphonic acid) ≥98,0% (durch titrimetrische Analyse), Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
N,N-Bis(phosphonomethyl)glycine, N-(Carboxymethyl)iminodi(methylphosphinic acid), Glyphosine
N,N-Bis(phosphonomethyl)glycine is present in the ligand bound to human apotransferrin.
Used as a complexing agent, pK-values: pK1: ·2, pK2: 2.0, pK3: 5.20, pK4: 6.77, pK5: 10.89.
Glyphosine binds to a pocket of MHC class II molecules I-Ag7 and DQ8, and modify T cell responses to the autoantigenic insulin B chain fragment 9–23 (B:9–23) peptide. Glyphosine stimulate IL-10 production and delay onset of diabetes in non-obese NOD mice.†
Formel:
C₄H₁₁NO₈P₂ Molecular Weight: 263,08 g/mol |
MDL:
MFCD00056670 CAS-Nummer: 2439-99-8 EINECS: 219-468-4 UN: 3261 ADR: 8,III |
Spezifikation Testergebnisse
Assay | ≥98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...