Arachidonic Acid 1,2,3,4,5-¹³C ≥95%
Lieferant: Cayman Chemical
|
Gefahr
|
Synonyme:
5Z,8Z,11Z,14Z-eicosatetraenoic-1,2,3,4,5-13C5 acid, AA 1,2,3,4,5-¹³C
Arachidonic acid (Item No. 90010) is an essential fatty acid and a precursor for all prostaglandins, thromboxanes, and leukotrienes.
- A solution in methanol
Virtually all cellular arachidonic acid is esterified in membrane phospholipids where its presence is tightly regulated through multiple interconnected pathways. Free arachidonic acid is a transient, critical substrate for the biosynthesis of eicosanoid second messengers. Receptor-stimulated release, metabolism, and re-uptake of free arachidonate are all important aspects of cell signaling and inflammation. Arachidonic acid 1,2,3,4,5-13C is an isotopically enriched form of this PUFA with carbon-13 occurring at positions 1, 2, 3, 4, and 5. It can be used to study metabolic processes related to arachidonic acid by means of mass spectrometry.
3 (6.1)
Formel:
C₁₅[¹³C]₅H₃₂O₂ Molecular Weight: 309,4 g/mol Lagertemperatur: Gefrierschrank |
UN:
1230 ADR: 3 (6.1),II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...