Eusolex® HMS

Lieferant: Merck

1.11412.1000 1.11412.9200_U 1.11412.9025_U 1.11412.9025 1.11412.9200 1.11412.9201
1.11412.1000EA 0 EUR
1.11412.1000 MERC1.11412.9200_U MERC1.11412.9025_U 1.11412.9025 1.11412.9200 1.11412.9201
Eusolex® HMS
Eusolex®

Organic UV Filter


Eusolex® HMS is a clear, liquid UVB filter, oil soluble with excellent dissolving properties for solid UV filters.


INCI name: Homosalate

Formel: C₁₆H₂₂O₃
Dichte: 1,05 g/cm3 (25 °C)
Flammpunkt: 90 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 118-56-9

Bestellen

Spezifikation Testergebnisse

Color colourless to yellowish
Appearance clear viscous liquid
Odour weak odour
Assay Homosalate (GC; sum of the two isomeric compounds) ≥ 98 % (a/a)
Assay Homosalate (GC) 90 - 110 % (w/w)
Assay via absorbance ≥ 98 %
Methyl salicylate (GC) ≤ 100 ppm
Identity (IR-spectrum) passes test
Identity (UV-spectrum) passes test
Refractive index (n 20/D) 1.516-1.519
Specific Density (d25/25) 1.049 - 1.053
Specific absorbance (306 nm; 1%; 1cm; ethanol) 170- 180
Heavy metals (as Pb) ≤ 0.002 %
As (Arsenic) ≤ 0.0002 %
Residual solvent Methanol (HS-GC) ≤ 2000 ppm
Salicylic acid passes test
Microbial Purity (Total Viable Aerobic Count) ≤ 10² CFU/g
E.coli absent in 1 g
Pseudomonas aeruginosa absent in 1 g
Staphylococcus aureus absent in 1 g
Candida albicans absent in 1 g
Salmonella species absent in 10 g
We confirm that the product meets the purity requirements defined in USP und JSQI by applying our internal testing methods which might deviate from those under USP und JSQI.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR