DNS Natriumsalz (Desoxyribonukleinsäure Natriumsalz) (aus Kalbsthymus), Calbiochem®, Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
Thymonucleic acid sodium salt, DNA sodium salt
2618-1
2618-250
2618-1EA
412
EUR
2618-1
2618-250
DNS Natriumsalz (Desoxyribonukleinsäure Natriumsalz) (aus Kalbsthymus), Calbiochem®, Millipore®
DNA sodium salt (Deoxyribonucleic acid sodium salt)
It is a purified DNA from calf thymus and an essential component of chromosomes
Special Instructions: Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 year at -20°C.
Lagertemperatur:
Gefrierschrank |
MDL:
MFCD00130921 CAS-Nummer: 9007-49-2 |
Spezifikation Testergebnisse
hyperchromicity | ≥27% |
S20w | 15.4-17.4 |
A260/280 nm | ≥ 1.8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...