Dexmedetomidine hydrochloride ≥99%

Lieferant: Cayman Chemical
Achtung

Synonyme: Dex, (S)-Medetomidine hydrochloride, (+)-Medetomidine hydrochloride, (S)-4-[1-(2,3-Dimethylphenyl)ethyl]-1H-imidazole hydrochloride, (S)-4-(α,2,3-Trimethybenzyl)-1H-imidazole hydrochloride, Precedex, 5-[(1S)-1-(2,3-Dimethylphenyl)ethyl]-1H-imidazole hydrochloride

15581-50 15581-5
CAYM15581-50EA 349 EUR
CAYM15581-50 CAYM15581-5
Dexmedetomidine hydrochloride ≥99%
Dexmedetomidine hydrochloride

Dexmedetomidine is the (S)-enantiomer of medetomidine, an imidazole compound that binds to α2-adrenoceptors (Ki = 1.08 nM) with high affinity and selectivity compared to that of α1-adrenoceptors (Ki = 1.8 μM). Dexmedetomidine produces analgesic, sedative, and anxiolytic effects through both presynaptic activation of α2-adrenoceptors, which inhibits norepinephrine release, and postsynaptic activation of α2-adrenoceptors, which inhibits sympathetic activity and decreases blood pressure and heart rate.


  • Formulation : A white to off-white solid
  • λmax: 214 nm

Formel: C₁₃H₁₆N₂·HCl
Molecular Weight: 236,74 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12912677
CAS-Nummer: 145108-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR