Methyl-β-cyclodextrin (mixture of several Methylated), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methylated-β-Cyclodextrin, Methyl-β-cyclodextrin

779873-100G 779873-10KG 779873-1KG 779873-500G
SIAL779873-100GEA 483 EUR
SIAL779873-100G SIAL779873-10KG SIAL779873-1KG SIAL779873-500G
Methyl-β-cyclodextrin (mixture of several Methylated), Sigma-Aldrich®
Methyl-beta-cyclodextrin (mixture of several Methylated)

Methyl-β-cyclodextrin is a macrocyclic compound that can form inclusion complexes with a number of guest molecules. It shows higher solubility in aqueous solutions and greater solubilising and complexing power compared to the parent β-cyclodextrin.


MβCD can be used to enhance the enzyme activity and enantioselectivity of subtilisin enzyme in organic solvents.


Produced by Wacker Chemie AG.

Molecular Weight: 1303,3000 g/mol
Flammpunkt: 187 °C
MDL: MFCD00074980
CAS-Nummer: 128446-36-6

Bestellen

Spezifikation Testergebnisse

Assay ≥98.0% Cyclodextrin Basis
Appearance (Colour) White to Off White
Appearance (Form) Powder or Crystals
Halides (as Chloride) ≤0.20%
Specific Rotation (20/D) 154.0 - 165.0 Degrees
Loss on Drying ≤5.0%
Residue on Ignition ≤0.50%
Infrared Spectrum Conforms to Structure
UV Method 10% in H₂O
Reducing Substances ≤0.50%
Volatile Impurities Volatile organics ≤50 ppm
Absorbance, 220 - 350 nm ≤1.0
Absorbance, 350 - 650 nm ≤0.10
Degree of Substitution Molar Substitution 1.7 - 1.9
Heavy Metals ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR