Aluminum phosphate, Reagenzqualität, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: Aluminium phosphate

341452-100G 341452-500G
SIAL341452-100GEA 64.6 EUR
SIAL341452-100G SIAL341452-500G
Aluminum phosphate, Reagenzqualität, Sigma-Aldrich®
Aluminum phosphate
Formel: AlPO₄
Molecular Weight: 121.951 g/mol
Dichte: 2.56 g/ml (25 °C) (Lit.)
MDL: MFCD00003430
CAS-Nummer: 7784-30-7
EINECS: 232-056-9
UN: 3260
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

Appearance White to Off-White Powder
Loss on Ignition Typically <1%
ICP Assay Confirms Aluminum And Phosphorus Components


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR