6-OAU ≥98%

Lieferant: Cayman Chemical

Synonyme: 6-(Octylamino)pyrimidine-2,4(1H,3H)-dione, 6-(octylamino)-2,4(1H,3H)-pyrimidinedione

17687-1 17687-10 17687-5
CAYM17687-1EA 124 EUR
CAYM17687-1 CAYM17687-10 CAYM17687-5
6-OAU ≥98%
6-OAU

GPR84 is a Gαi protein-coupled receptor that is activated by medium-chain free fatty acids, but only at micromolar concentrations. GPR84 is expressed in leukocytes and its expression is further induced by LPS.


  • Formulation: A crystalline solid
  • λmax: 265 nm
  • Stability: 2 years at -20°C


6-OAU is a synthetic agonist for GPR84 (EC50 = 512 nM). It has insignificant actions at a panel of 69 other receptors, channels, or transporters. 6-OAU promotes chemotaxis in human neutrophils (EC50 = 318 nM) and amplifies TNF-α production by LPS-treated U937 macrophages. In rats, 6-OAU (1 mg/ml) drives the recruitment of neutrophils and macrophages when injected into a subcutaneous air pouch and raises serum levels of CXCL1 (growth-regulated α protein) when delivered intravenously.

Formel: C₁₂H₂₁N₃O₂
Molecular Weight: 239,30 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD28900678
CAS-Nummer: 83797-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR