Indo-1 AM ≥96% (durch HPLC) fluoreszierender Indikator, zur Kalziumbestimmung
Lieferant: Biotium
Synonyme: 1-[2-Amino-5-(6-carboxyindol-2-yl)phenoxy]-2-(2'-amino-5'-methylphenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethylester, Indo-1-(acetoxymethyl) ester, 4-(6-Carboxy-2-indolyl)-4′-methyl-2,2′-(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, Indo-1 AM Ester
50044 50043 50043-1
50044.EA 293 EUR
50044. 50043. 50043-1
Indo-1 AM ≥96% (durch HPLC) fluoreszierender Indikator, zur Kalziumbestimmung
Indo-1 AM
Indo-1, AM ester is membrane-permeant and thus can be loaded into cells via incubation. Because of the relatively low water solubility of the AM ester, Pluronic F-127, a mild detergent, is often used as a dispersing agent to facilitate cell loading. Indo-1-AM Ester itself does not bind Ca2+ , but it is readily hydrolyzed to indo-1 by endogenous esterases once it enters cells.
  • Absorption/Emission 356 nm/478 nm (no Ca2+); 330 nm/400 nm (high Ca2+)
  • Extinction Coefficient 33,000 M -1 cm -1 (no Ca2+); 33,000 M -1 cm -1 (high Ca2+)
  • Cell permeability: Membrane permeant
  • Light yellow solid soluble in DMSO
  • Store desiccated at -20°C and protect from light, especially in solution
Formel: C₄₇H₅₁N₃O₂₂
Molecular Weight: 1009.91 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00036955
CAS-Nummer: 112926-02-0