Thallium(I)bromid, wasserfrei ≥99,999%, Pulver, Optipur®

Lieferant: Merck

Synonyme: Thalliumbromid

1.89356.0100 1.89356.1000
1.89356.0100EA 0 EUR
1.89356.0100 1.89356.1000
Thallium(I)bromid, wasserfrei ≥99,999%, Pulver, Optipur®
Thallium(I)bromid

Optipur® materials are inorganic compounds, primarily metal halides, and available as anhydrous powders, beads or pieces for immediate usability and easy handling. They are of the highest purity, and specifically suited for the production of single crystals, used in demanding optical applications or scintillators, for example medical imaging and radiation detection.


  • High purity
  • Anhydrous powders or beads
  • Mainly metal halides
  • Immediate usability and easy handling
  • Special packaging for hygroscopic materials


The purity of each production batch is strictly controlled with careful attention paid to impurities that have an influence on transmission, light scattering, or scintillation efficiency. Hygroscopic materials are typically vacuum-dried at high temperature and are packaged for dry storage.

Formel: BrTl
Molecular Weight: 284,29 g/mol
MDL: MFCD00011273
CAS-Nummer: 7789-40-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR