2-(Diisopropylamino)ethyl methacrylate 97% inhibiert, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: 2-(Diisopropylamino)ethyl methacrylate

Stabilisierung: Stabilised with 4-methoxyphenol (MEHQ) ~100 ppm

730971-25G 730971-100G
SIAL730971-25GEA 111 EUR
SIAL730971-25G SIAL730971-100G
2-(Diisopropylamino)ethyl methacrylate 97% inhibiert, Sigma-Aldrich®
2-(Diisopropylamino)ethyl methacrylate

2-(Diisopropylamino)ethyl methacrylate, Stabilized with 80-100 ppm MEHQ

Formel: C₁₂H₂₃NO₂
Molecular Weight: 213,32 g/mol
Dichte: 0,900 g/cm³ (25 °C)
Flammpunkt: 98 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00072647
CAS-Nummer: 16715-83-6
EINECS: 240-772-8

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Colorless to Tan
Appearance (Form) Liquid
Proton NMR spectrum Conforms to Structure
Purity (GC) ≥96,5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR