Green CMFDA ≥95% fluoreszierender Farbstoff

Lieferant: Cayman Chemical

Synonyme: 3',6'-bis(acetyloxy)-5-(chloromethyl)-spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 5-(Chloromethyl)-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-diyl diacetate, 5-Chloromethylfluorescein diacetate

19583-1 19583-5 19583-10
CAYM19583-1EA 187 EUR
CAYM19583-1 CAYM19583-5 CAYM19583-10
Green CMFDA ≥95% fluoreszierender Farbstoff
Green CMFDA

Green CMFDA is a cell-permeable fluorescent probe that freely enters living cells, where it is transformed into a cell-impermeable, soluble, fluorescent compound. The resulting molecule does not affect cell viability or proliferation and stably fluoresces for at least 72 hours, allowing for its use in cell tracking. It is also transferred to daughter cells.


  • Emission 517 nm
  • Excitation 492 nm
  • Formulation A crystalline solid
  • λmax 220, 291 nm


Green CMFDA is brightly fluorescent with standard filters (ex/em max = 492/517 nm) and is often used with additional fluorescent dyes and proteins.

Formel: C₂₅H₁₇ClO₇
Molecular Weight: 464,9 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00467170
CAS-Nummer: 136832-63-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR