5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE)
Lieferant: Apollo Scientific
Synonyme: NHS-Fluorescein, FLUOS, 5(6)-FAM SE
BIC1074-100MG BIC1074-1G
APOSBIC1074-100MGEA 174 EUR
APOSBIC1074-100MG APOSBIC1074-1G
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE)
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE)
Formel: C₂₅H₁₅NO₉
Molecular Weight: 473,39 g/mol
MDL: MFCD04119713
CAS-Nummer: 117548-22-8