Patulin ≥98%, kristalliner Feststoff
Lieferant: Cayman Chemical
|
Gefahr
|
11346-5
11346-10
CAYM11346-5EA
96
EUR
CAYM11346-5
CAYM11346-10
Patulin ≥98%, kristalliner Feststoff
Patulin
Patulin is a mycotoxin produced by a variety of molds commonly found in rotting apples, including Aspergillus and Penicillium. Patulin-induced nephropathy and gastrointestinal tract malfunction have been demonstrated in several animal models.
Formel:
C₇H₆O₄ Molecular Weight: 154,121 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00005858 CAS-Nummer: 149-29-1 UN: 3462 ADR: 6.1,II |
Spezifikation Testergebnisse
Purity | ≥98% |
Formulation | A Crystalline Solid |
λmax | 275 nm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...