Treprostinil ≥98%

Lieferant: Cayman Chemical
Gefahr

Synonyme: Uniprost, U-62840, LRX 15, Rumodolin, 2-(((1R,2R,3aS,9aS)-2-Hydroxy-1-((S)-3-hydroxyoctyl)-2,3,3a,4,9,9a-hexahydro-1H-cyclopenta[b]naphthalen-5-yl)oxy)acetic acid, BW 15AU

10162-1 10162-10 10162-5
CAYM10162-1EA 129 EUR
CAYM10162-1 CAYM10162-10 CAYM10162-5
Treprostinil ≥98%
Treprostinil

Treprostinil is a stable analog of prostacyclin that is used clinically for the treatment of PPH under the trade name Remodulin™. The structural modifications in treprostinil compared to prostacyclin increase the plasma half-life from 2 minutes to 34 and 85 minutes for intravenous and subcutaneous infusion of the drug, respectively


Bitte beachten Warning - this product is not for human or veterinary use.

Formel: C₂₃H₃₄O₅
Lagertemperatur: Gefrierschrank
MDL: MFCD00888847
CAS-Nummer: 81846-19-7

Bestellen

Spezifikation Testergebnisse

Purity ≥98%
Formulation A Crystalline Solid
λmax 206, 218, 277 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR