1,1,1,3,3,3-Hexamethyldisilazane : Chlorotrimethylsilane (HMDS + TMCS)-Lösung (10 vol parts) in Pyridin , Silylating mixture I according to Sweeley, LiChropur™ zur GC-Derivatisierung, Supelco®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ100
85431-10ML
85431-10X1ML
85431-25ML
85431-10MLEA
116
EUR
85431-10ML
85431-10X1ML
85431-25ML
1,1,1,3,3,3-Hexamethyldisilazane : Chlorotrimethylsilane (HMDS + TMCS)-Lösung (10 vol parts) in Pyridin , Silylating mixture I according to Sweeley, LiChropur™ zur GC-Derivatisierung, Supelco®
1,1,1,3,3,3-Hexamethyldisilazane : Chlorotrimethylsilane (HMDS + TMCS)
Hexamethyldisilazane/chlorotrimethylsilane/pyridine = 2:1:10 (v/v/v), useful silylating mixture for the silylation of hydroxy-compounds.
Flammpunkt:
17 °C Lagertemperatur: Kühlschrank |
UN:
1992 ADR: 3 (6.1),II |