Benfotiamine ≥98%
Lieferant: Cayman Chemical
Synonyme:
S-(2-(N-((4-Amino-2-methylpyrimidin-5-yl)methyl)formamido)-5-(phosphonooxy)pent-2-en-3-yl) benzothioate, S-Benzoylthiamine O-Monophosphate
Benfotiamine is a lipid-soluble form of vitamin B1 (thiamine). in vitro, it corrects defective replication of, and prevents formation of advanced glycosylation end products (AGEs) in, human umbilical vein endothelial cells (HUVECs) grown under high glucose conditions. in vivo, administration of benfotiamine increases nerve conduction velocity (NCV) and prevents microalbuminuria, proteinuria, and formation of AGEs in mice with streptozotocin-induced diabetes.
Benfotiamine reduces liver levels of aspartate and alanine aminotransferases, markers of hepatic damage, and lipid peroxidation in a rat model of acute ethanol intoxication. Administration of benfotiamine reduces the number of amyloid plaques and amount of phosphorylated tau in a transgenic mouse model of alzheimer's disease. It also improves spatial memory performance in the morris water maze.
Formel:
C₁₉H₂₃N₄O₆PS Molecular Weight: 466,5 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00057343 CAS-Nummer: 22457-89-2 |
Spezifikation Testergebnisse
Purity | ≥98% |
Formulation | A Crystalline Solid |
λmax | 243 nm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...