1,1,1,3,3,3-Hexafluor-2-propanol 99,8, LiChropur™ derivatisation_GC, Supelco®
Lieferant: Merck
Gefahr
Synonyme: 1,1,1,3,3,3-Hexafluorisopropanol, HFIP, 1,1,1,3,3,3-Hexafluor-2-hydroxypropan
52517-10ML 52517-50ML 52517-10X1ML
52517-10MLEA 247 EUR
52517-10ML 52517-50ML 52517-10X1ML
1,1,1,3,3,3-Hexafluor-2-propanol 99,8, LiChropur™ derivatisation_GC, Supelco®
1,1,1,3,3,3-Hexafluor-2-propanol
Hexafluoroisopropanol (HFP) is generally used to denature the native state of proteins, and it also stabilizes the a-helical conformation in unfolded peptides and proteins like ß-lactoglobulin and melittin
Suitable for the derivatization of glutamic acid, homovanillic, iso-homovanillic and dihydroxyphenyl acetic acids and Υ-aminobutyric acid.
Formel: C₃H₂F₆O
Molecular Weight: 168.04 g/mol
Siedepunkt: 59 °C (Lit.)
Schmelzpunkt: -96
MDL: MFCD00011651
CAS-Nummer: 920-66-1
UN: 3265
ADR: 8 II
Merck Index: 13,09598
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5da99ea9af42756619fc0e4094bac155adbedf3a&vid=5da99ea9af42756619fc0e4094bac155adbedf3a&prid=web&clid=SAPDAM