di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP), Tabletten
Lieferant: Uptima
Synonyme:
pNPP, ortho-Phosphorsäure(4-nitrophenyl)ester Dinatriumsalz Hexahydrat, 4-Nitrophenylphosphat Dinatriumsalz Hexahydrat, p-Nitrophenylphosphat Dinatriumsalz Hexahydrat, ortho-Phosphorsäuremono(4-nitrophenyl)ester Dinatriumsalz Hexahydrat, Phosphatasesubstrat, 4-Nitrophenyldihydrogenphosphat Dinatriumsalz Hexahydrat
UP89562F
UP89562G
UPTIUP89562FEA
0
EUR
UPTIUP89562F
UPTIUP89562G
di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP), Tabletten
di-Natrium-4-nitrophenylphosphat Hexahydrat (pNPP)
Formel:
C₆H₄NNa₂O₆P·6H₂O Molecular Weight: 371.144 g/mol Schmelzpunkt: 300 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00066288 CAS-Nummer: 4264-83-9 EINECS: 224-246-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...