MCI GEL® CHP20P 75 - 150 μm, Supelco®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
13629-U
13630-U
13629-UEA
279
EUR
13629-U
13630-U
MCI GEL® CHP20P 75 - 150 μm, Supelco®
MCI GEL® CHP20P
MCI GEL® CHP20P is a highly porous styrene-divinylbenzene polymer resin used as column packing material.
- Surface area: ~500 m²/g
Polyaromatic adsorbent resin designed for biopharmaceutical separations: aromatic compounds, peptides, steroids, desalting, etc. Very good for reversed phase chromatography applications, even with nonaqueous solvents.