2′,3′-Dideoxycytidine 5′-triphosphate sodium salt ≥90% (durch HPLC), Sigma-Aldrich®

Lieferant: Merck

Synonyme: ddCTP

D7159-1MG D7159-2MG
SIALD7159-1MGEA 448 EUR
SIALD7159-1MG SIALD7159-2MG
2′,3′-Dideoxycytidine 5′-triphosphate sodium salt ≥90% (durch HPLC), Sigma-Aldrich®
2′,3′-Dideoxycytidine 5′-triphosphate sodium salt

Inhibitor of DNA polymerase I-catalyzed chain elongation.


Complexes of ddCTP with DNA polymerase β were studied by x-ray crystallography, with implications for inhibitory activity against HIV reverse transcriptase.

Formel: C₉H₁₆N₃O₁₂P₃
Molecular Weight: 451,16 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00717536
CAS-Nummer: 132619-66-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR