Calcium ionophore IV, Selectophore™, Funktion getestet Ionophor, Supelco®
Lieferant: Merck
Synonyme: N,N-Dicyclohexyl-2-(2-(dioctadecylamino)-2-oxoethoxy)acetamide, N,N-Dicyclohexyl-N′,N′-dioctadecyl-3-oxapentanediamide, ETH 5234, N,N-Dicyclohexyl-N′,N′-dioctadecyl-diglycolic diamide
M-Clarity™ Qualitätsstufe = MQ100
21198-250MG-F 21198-50MG-F
21198-250MG-FEA 1080 EUR
21198-250MG-F 21198-50MG-F
Calcium ionophore IV, Selectophore™, Funktion getestet Ionophor, Supelco®
Calcium ionophore IV
Calcium ionophore IV (ETH 5234) was used in preparing membrane for the pseudoreference electrode in tetrahydrofuran, in a study to demonstrate electrochemical sample matrix elimination for trace level potentiometric detection.
Calcium ionophore IV may be used in the preparation of Ca2+-selective membrane utilizing AlGaN/GaN transistors for potentiometric calcium ion detection.
Extremely lipophilic Ca2+-selective ionophore for chemical sensors of long lifetime; it induces selectivities for Ca over Mg, Na and K of 104, 106, and 108, respectively
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
Formel: C₅₂H₁₀₀N₂O₃
Molecular Weight: 801,36 g/mol
MDL: MFCD00213934
CAS-Nummer: 126572-74-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=54020b8ca0f03184560c41dd317e611949509db0&vid=ae329d6ab8822df2ef182bde76412417017e5b52&prid=web&clid=SAPDAM