Chloride ionophore III, Selectophore™ Ionophor, Supelco®
Lieferant: Merck
|
Gefahr
|
Synonyme:
3,6-Didodecyloxy-4,5-dimethyl-o-phenylene-bis(mercury chloride), ETH 9033
M-Clarity™ Qualitätsstufe = MQ100
Chloride selective ionophore for Cl--electrodes. ETH 9033 (chromoionophore III) may be used to monitor Cl- activity during the development of chloride sensitive probes encapsulated by biologically localized embedding.
It may also be used with Cl- ion sensitive ISFETs (ion-sensitive field-effect transistor) with photocured polyurethane-based polymer membrane to study sodium chloride solutions and to determine the chloride ions.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
Formel:
C₃₂H₅₆Cl₂Hg₂O₂ Molecular Weight: 944.87 g/mol |
MDL:
MFCD00676703 CAS-Nummer: 178959-28-9 UN: 2025 ADR: 6.1 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...