Copper(II) ionophore I, Selectophore™ Ionophor, Supelco®
Lieferant: Merck
Synonyme: o-Xylylenebis(N,N-diisobutyldithiocarbamate), o-XBDiBDTC
61193-50MG
61193-50MGEA 274 EUR
61193-50MG
Copper(II) ionophore I, Selectophore™ Ionophor, Supelco®
Copper(II) ionophore I
Neutral ionophore for Cu2+-selective polymer membrane electrodes
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
Formel: C₂₆H₄₄N₂S₄
Molecular Weight: 512.90 g/mol
MDL: MFCD00214160
CAS-Nummer: 125769-67-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff1937fb0eff4e411c3b5d68ad8c8ab9ff9c8f47&vid=4e53b8fe48cc2e9f6628e1d9f3d3a84ff37429a4&prid=web&clid=SAPDAM