(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥99,0%, LiChropur™ zur chiralen Derivatisierung, Supelco®
Lieferant: Merck
Gefahr
Synonyme: (+)-Mosher's Acid Chloride, (+)-MTPA-Cl, (S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, (S)-3,3,3-Trifluoro-2-methoxy-2-phenylpropanoyl chloride
M-Clarity™ Qualitätsstufe = MQ100
65365-100MG-F 65365-500MG-F
65365-100MG-FEA 129 EUR
65365-100MG-F 65365-500MG-F
(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥99,0%, LiChropur™ zur chiralen Derivatisierung, Supelco®
(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
(R)-(-)-a-Methoxy-a-(trifluoromethyl)phenylacetyl chloride (Mosher’s acid chloride) is generally used as a chiral derivatizing agent.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
Formel: C₁₀H₈ClF₃O₂
Molecular Weight: 252,62 g/mol
MDL: MFCD00067105
CAS-Nummer: 20445-33-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f52c12e355b4497707c2905723aa09c047497022&vid=69da0e9ae264d2748152dd3dbf0014a243600059&prid=web&clid=SAPDAM