Dimethyldichlorsilan 5% in Heptan Silanization solution I, Selectophore™ Ionophor, Supelco®
Lieferant: Merck
Gefahr
Synonyme: Dichlor(dimethyl)silan, Dichlordimethylsilan, Dimethylsilyldichlorid
M-Clarity™ Qualitätsstufe = MQ100
85126-1L 85126-250ML
85126-1LEA 208 EUR
85126-1L 85126-250ML
Dimethyldichlorsilan 5% in Heptan Silanization solution I, Selectophore™ Ionophor, Supelco®
Dimethyldichlorsilan
Ready-to-use solution for silanizing micro-electrodes.
Formel: C₂H₆Cl₂Si
Molecular Weight: 129.06 g/mol
Siedepunkt: 70
Schmelzpunkt: -76
Flammpunkt: -16
MDL: MFCD00000491
CAS-Nummer: 75-78-5
UN: 2924
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d29a64e31328c65f3e1562b829e50639e996c41d&vid=0a4419ea13f3484b5717c588fe35e543f9fcbec9&prid=web&clid=SAPDAM