BrettPhos palladacycle 3rd generation 98%
Lieferant: Thermo Scientific
Achtung
Synonyme: [(2-Di-cyclohexylphosphino-3,6-dimethoxy-2′,4′,6′- triisopropyl-1,1′-biphenyl)-2-(2′-amino-1,1′ -biphenyl)]palladium(II) methanesulphonate, BrettPhos Pd G3
47250.03 47250.MD
47250.03EA 405 EUR
47250.03 47250.MD
BrettPhos palladacycle 3rd generation 98%
BrettPhos palladacycle 3rd generation
Formel: C₄₈H₆₆NO₅PPdS
Molecular Weight: 906,50 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22572666
CAS-Nummer: 1470372-59-8
Bestellen
Spezifikation Testergebnisse
Appearance (Color) Yellow
Form Powder
Elemental Analysis 62,0 to 65,0 wt% Carbon