Tetrahydrofuran 99,7 Unstabilised, AcroSeal®, Spectrophotometric grade
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: Tetramethylenoxid, Furantetrahydrid, 1,4-Oxidobutan, Oxolan, THF
Argon_filled
47329.AK 47329.K2 47329.M1
47329.AKEA 67.5 EUR
47329.AK 47329.K2 47329.M1
Tetrahydrofuran 99,7 Unstabilised, AcroSeal®, Spectrophotometric grade
Tetrahydrofuran
AcroSeal™ septum bottled solvent.
  • Supports optimum performance of your moisture-sensitive work
Formel: C₄H₈O
Molecular Weight: 72.11 g/mol
Siedepunkt: 66 °C
Schmelzpunkt: –108 °C
Dichte: 0.889
Flammpunkt: –17°C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005356
CAS-Nummer: 109-99-9
EINECS: 203-726-8
UN: 2056
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM