2-[Ethyl(phenyl)amino]ethanol
Lieferant: ABCR
Synonyme:
N-Ethyl-N-(2-hydroxyethyl)aniline, Ethylphenylethanolamine, 2-(Ethyl(phenyl)amino)ethanol, N-Ethyl-N-2-ethanolaniline
TCE0062.0200
TCE0062.0500
ABCRTCE0062.0200EA
0
EUR
ABCRTCE0062.0200
ABCRTCE0062.0500
2-[Ethyl(phenyl)amino]ethanol
2-(N-Ethylanilino)ethanol
Formel:
C₁₀H₁₅NO Molecular Weight: 165,24 g/mol |
MDL:
MFCD00020575 CAS-Nummer: 92-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...