Picolinoylchlorid Hydrochlorid
Lieferant: ABCR
Synonyme:
Pyridine-2-carbonyl chloride hydrochloride
TCP0619.0005
TCP0619.0025
ABCRTCP0619.0005EA
0
EUR
ABCRTCP0619.0005
ABCRTCP0619.0025
Picolinoylchlorid Hydrochlorid
Picolinoylchlorid Hydrochlorid
Formel:
C₆H₅Cl₂NO Molecular Weight: 178,0160 g/mol |
MDL:
MFCD00060193 CAS-Nummer: 39901-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...