Cell culture media, Minimum Essential Medium (MEM)
Lieferant: PAN BIOTECH
P04-08050 P04-08509 P04-08500 P04-09500 P04-10500
PANBP04-08050EA 15.3 EUR
PANBP04-08050 PANBP04-08509 PANBP04-08500 PANBP04-09500 PANBP04-10500
Cell culture media, Minimum Essential Medium (MEM)
Zellkulturmedien
MEM is an advancement of the BME and the base medium of many further modifications. Because BME did not fulfil all requirements for some mammalian and HeLa cells, a better variation had to be developed. Today, MEM is one of the most used synthetic media and shows its versatility by supplementing with amino acids including Hank's or Earle's salts. Even the addition of only small amounts of FBS results in a positive effect on cell growth.
  • Store from 2 to 8 °C