Kieselsäure Kaliumsalz, wasserfrei
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: Kaliumsilicat
44493.22 44493.36 44493.A3
44493.22EA 33.5 EUR
44493.22 44493.36 44493.A3
Kieselsäure Kaliumsalz, wasserfrei
Silicic acid potassium salt
Potassium silicate is used in binders, carbon electrodes, lead pencils, protective coatings, insoluble pigments, phosphors on television tubes and detergent additives. It acts as a catalyst to make glass, ceramics, welding rods, adhesives, cements, gels, paints, pulp and paper production. Further, it is used in horticulture as a soluble source of potassium and silicon. It acts as a corrosion inhibitor.
Bitte beachten Moisture sensitive. Incompatible with strong oxidizing agents.
Lagertemperatur: Raumtemperatur
MDL: MFCD00145152
CAS-Nummer: 1312-76-1
EINECS: 215-199-1
Merck Index: 14,7668
Bestellen
Spezifikation Testergebnisse
Assay (unspecified) 28,0 - 28,8% K₂O
TECHNISCHE DATEN
Environmentally Preferable