Glycerin 87% ≥87%, reinst
Lieferant: Bernd Kraft
Synonyme:
1,2,3-Propantriol, Glycerol
07090.3000
07090.5000
07090.7000
07090.2000
07090.4000
KRAF07090.3000EA
55.4
EUR
KRAF07090.3000
KRAF07090.5000
KRAF07090.7000
KRAF07090.2000
KRAF07090.4000
Glycerin 87% ≥87%, reinst
Glycerin
Formel:
HOCH₂CH(OH)CH₂OH Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004722 CAS-Nummer: 56-81-5 EINECS: 200-289-5 Merck Index: 13,04497 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...