Rifampicin
Lieferant: Serva
Synonyme:
5,6,9,17,19,21-Hexahydroxy-23-methoxy- 2,4,12,16,18,20,22-heptamethyl-8-[N-(4-methyl- 1-piperazinyl)formimidoyl]-2,7-(epoxypentadeca- [1,11,13]trienimino)-naphtho[2,1-b]furan- 1,11(2H)-dion-21-acetat, Rifampin
34514.01
34514.02
SERA34514.01EA
79.7
EUR
SERA34514.01
SERA34514.02
Rifampicin
Rifampicin
Formel:
C₄₃H₅₈N₄O₁₂ Molecular Weight: 822.95 g/mol Schmelzpunkt: 183…188 °C |
MDL:
MFCD00151389 CAS-Nummer: 13292-46-1 EINECS: 236-312-0 Merck Index: 12,08382 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...