Orcein, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Naturrot 28

MQ100

O7380-10G O7380-5G
O7380-10GEA 194 EUR
O7380-10G O7380-5G
Orcein, Sigma-Aldrich®
Orcein
MDL: MFCD00062310
CAS-Nummer: 1400-62-0
EINECS: 215-750-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR